Learn More
Andwin Scientific BLOTT PAPER 7X10 100/PK
SDP
Supplier:  Andwin Scientific 28298030
Encompass_Preferred
Blotting Paper, Grade 703, Size 7 x 10cm, Weight 185g m2, Thick 0.38mm, 100mL at 10cm Head of water, Klemm wicking rate test 626 sec., 7.5cm rise, Made of 100 cotton fiber, Uniform and smooth, Produced using ultrapure water
Catalog No. NC9093750