Learn More
Andwin Scientific VWR BLUE DISP SPATULA/PK/300
SDP
Catalog No. NC9996854
Encompass_Preferred
The disposable spatulas features a unique double-ended design that allows this product to be used as a spoon, scoop, scraper, stirring rod, or policeman. This product is disposable and economical. It can be used for transferring, manipulating, and mixing materials and can also reduce the chance of cross-contamination. This product is sold in a case of 300 blue spatulas. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc
Catalog No. NC9996854 Supplier Andwin Scientific Supplier No. 80081188
The disposable spatulas features a unique double-ended design that allows this product to be used as a spoon, scoop, scraper, stirring rod, or policeman. This product is disposable and economical. It can be used for transferring, manipulating, and mixing materials and can also reduce the chance of cross-contamination. This product is sold in a case of 300 blue spatulas. The V design at the neck of the disposable laboratory spatula is a trademark of LevGo, Inc