Learn More
Andwin Scientific VWR BRAND TUBES 1.5M PK/500
SDP
Catalog No. 5089990311
Encompass_Preferred
The disposable 1.5 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in a pack of 500.
Catalog No. 50-899-90311 Supplier Andwin Scientific Supplier No. 89000028
Add to Cart
Add to Cart
The disposable 1.5 mL microcentrifuge tubes are autoclavable, nonsterile, and graduated tubes that ensure easy and convenient sample measurement. This product also have flat caps and frosted areas on the sides that allows users easy sample identification and labeling. The caps can be opened with no difficulty, but will remain tightly closed during the process of centrifuging. Syringes can also penetrate the caps. This product is sold in a pack of 500.