Learn More
Andwin Scientific COVERGLASS 24X50MM PK 1 OZ
SDP
Catalog No. NC0125754
Encompass_Preferred
The Rectangular Coverglass feature superior chemical resistance and flatness. This product is constructed with pure white borosilicate glass and does not have bubbles or scratches. The microscope cover glasses measure 24 x 50 mm (31/32 x 2").
Catalog No. NC0125754 Supplier Andwin Scientific Supplier No. 16004322
The Rectangular Coverglass feature superior chemical resistance and flatness. This product is constructed with pure white borosilicate glass and does not have bubbles or scratches. The microscope cover glasses measure 24 x 50 mm (31/32 x 2").