Learn More
Andwin Scientific Disposable Culture Tubes 500/box
SDP
Catalog No. NC0866611
Encompass_Preferred
The 25x150mm 60ml culture tubes feature Type 1, Class B borosilicate glass construction, which prevents pH change in laboratory tests. It has well-rounded bottoms, fire-polished tops free of rough edges, uniform dimensions and convenient dispensing packages that keep them free from dust and other particulate contamination. It is resistant to chemical attack and weathering, avoiding leach out that contaminates solutions. Each tube is heat-treated at temperatures exceeding 500C (932F) immediately before packing, eliminating contamination, increasing strength, and reducing breakage. Tubes exceed USP Type I specifications.
Catalog No. NC0866611 Supplier Andwin Scientific Supplier No. 47729586
The 25x150mm 60ml culture tubes feature Type 1, Class B borosilicate glass construction, which prevents pH change in laboratory tests. It has well-rounded bottoms, fire-polished tops free of rough edges, uniform dimensions and convenient dispensing packages that keep them free from dust and other particulate contamination. It is resistant to chemical attack and weathering, avoiding leach out that contaminates solutions. Each tube is heat-treated at temperatures exceeding 500C (932F) immediately before packing, eliminating contamination, increasing strength, and reducing breakage. Tubes exceed USP Type I specifications.