Learn More
Andwin Scientific VWR DISP PIPET RES 50ML 100/CS
SDP
Supplier:  Andwin Scientific 89094682
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 50 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 100 white sterile reservoirs.
Catalog No. NC0584336