Learn More
Andwin Scientific VWR DISP PIPET RES 50ML 100/CS
SDP
Catalog No. NC0584334
Encompass_Preferred
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 100 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 100 natural-colored sterile reservoirs.
Catalog No. NC0584334 Supplier Andwin Scientific Supplier No. 89094678
The disposable pipetting reservoirs are made for multichannel pipetting applications. This product is compatible with any type of pipettors that are up to 12-channel width. This polystyrene reservoir has a unique trough construction, graduation marks on interior walls, and pour-off spouts on each corner. It has a volume capacity of 100 mL and has a dimension of 14.6 x 8.3 x 3 cm. This product is sold in a case of 100 natural-colored sterile reservoirs.