Learn More
Andwin Scientific VWR FRIDGE/FREEZER THERMOMETER
SDP
Catalog No. NC0567405
Encompass_Preferred
The Refrigerator/Freezer Thermometer features single display, maximum/minimum temperature memory, high/low alarm, 1-30mm waterproof cable, magnetic back and ranges of 14-122 deg.F and-58 to 158 deg.F.
Catalog No. NC0567405 Supplier Andwin Scientific Supplier No. 82021172
The Refrigerator/Freezer Thermometer features single display, maximum/minimum temperature memory, high/low alarm, 1-30mm waterproof cable, magnetic back and ranges of 14-122 deg.F and-58 to 158 deg.F.