Learn More
Andwin Scientific LTS Compatible Pipet Tips, 0.1-20 uL, Pre-sterilized, 960/pk
SDP
Catalog No. NC1778211
Encompass_Preferred
LTS Compatible pipet tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS compatible pipettors.
Catalog No. NC1778211 Supplier Andwin Scientific Supplier No. 76322158
LTS Compatible pipet tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS compatible pipettors.