Learn More
Andwin Scientific MICRO TUBES 1.7ML 5000CS
SDP
Catalog No. NC0580790
Encompass_Preferred
The Microcentrifuge Tubes provide ideal clarity. The tubes are compatible with all standard rotos. Caps are attached onto the tubes, which offer a positive seal design that can resist repeated cap closures and will not pop open during boiling. The tu
Catalog No. NC0580790 Supplier Andwin Scientific Supplier No. 87003294
The Microcentrifuge Tubes provide ideal clarity. The tubes are compatible with all standard rotos. Caps are attached onto the tubes, which offer a positive seal design that can resist repeated cap closures and will not pop open during boiling. The tu