Learn More
Andwin Scientific VWR Mini Centrifuge, 115V
SDP
Catalog No. nc1665546
Encompass_Preferred
The VWR Mini Centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes. Capacity: 8×1.5/2.0 and 32×0.2 mL
Catalog No. NC1665546 Supplier Andwin Scientific Supplier No. 76269064
The VWR Mini Centrifuge is designed for quick spin-downs of microcentrifuge tubes and PCR strips and tubes. Capacity: 8×1.5/2.0 and 32×0.2 mL