Learn More
Andwin Scientific VWR PBS-T BUFFER POWDER 100/CS
SDP
Catalog No. NC3937758
Encompass_Preferred
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.Buffer powder quickly dissolves to make 1L of 1X solutionNo weighing or measuring of separate componentsNo pH adjustments requiredEconomical shipping, long shelf lifeQuality and consistency guaranteedVWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.
Catalog No. NC3937758 Supplier Andwin Scientific Supplier No. 76371736
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.Buffer powder quickly dissolves to make 1L of 1X solutionNo weighing or measuring of separate componentsNo pH adjustments requiredEconomical shipping, long shelf lifeQuality and consistency guaranteedVWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.