Learn More
Andwin Scientific VWR SAFESEAL .65ML NAT.TUBE PK
SDP
Catalog No. NC0074118
Encompass_Preferred
MulTI SafeSeal Microcentrifuge Tube 0.65mL polypropylene. Closure wont pop open during boiling. -80 to +121[degree]C. 0.2mL graduations. Certified RNase- DNase- human DNA- and PCR inhibitor-free. Autoclavable. RCF: 30000g. Natural.
Catalog No. NC0074118 Supplier Andwin Scientific Supplier No. 53550970PK1000
MulTI SafeSeal Microcentrifuge Tube 0.65mL polypropylene. Closure wont pop open during boiling. -80 to +121[degree]C. 0.2mL graduations. Certified RNase- DNase- human DNA- and PCR inhibitor-free. Autoclavable. RCF: 30000g. Natural.