Learn More
Andwin Scientific Superfrost Slides, 75×25 mm, White, 20/cs
SDP
Catalog No. NC9946519
Encompass_Preferred
The VWR Superfrost Plus Micro Slide features a special treatment that eliminates the need for additional adhesives or protein coatings, and improves tissue adherence in microwave stainings.
Catalog No. NC9946519 Supplier Andwin Scientific Supplier No. 48311703CS
The VWR Superfrost Plus Micro Slide features a special treatment that eliminates the need for additional adhesives or protein coatings, and improves tissue adherence in microwave stainings.