Learn More
Andwin Scientific SLIDE SPRFST 25X75MM 72/PK
SDP
Catalog No. NC9872137
Encompass_Preferred
The Superfrost Plus Micro Slide has a special treatment that removes the necessity of additional adhesives or protein coatings and upgrades the tissue adherence in microwave stainings. This product is perfect for frozen tissue sections and cytology
Catalog No. NC9872137 Supplier Andwin Scientific Supplier No. 48311703
The Superfrost Plus Micro Slide has a special treatment that removes the necessity of additional adhesives or protein coatings and upgrades the tissue adherence in microwave stainings. This product is perfect for frozen tissue sections and cytology