Learn More
Andwin Scientific VWR STAINING DISH WHITE
SDP
Catalog No. NC9418051
Encompass_Preferred
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The staining dish is constructed from polypropylene and is sold in the color of white.
Catalog No. NC9418051 Supplier Andwin Scientific Supplier No. 25608906
The Tissue-Tek Slide Staining Dish is a replacement for the Tissue-Tek Slide Staining Set, a system specially made for histological or cytological staining procedures. The staining dish is constructed from polypropylene and is sold in the color of white.