Learn More
Andwin Scientific VWR SUPERCLEAR SCREW-CAP MICRO
SDP
Catalog No. NC0127657
Encompass_Preferred
The 1.5ml SuperClear screwcap microtube feature a conical bottom, external graduations, universal screw threads and smooth surface for applying labels. It is constructed from ultra clear resin for easily viewing of contents. It includes attached natural low-profile caps with an ethylene propylene O-ring ensuring secure seal. They are compatible with most common rotors, autoclavable at 251F, freezable to -130F and radiation sterilized.
Catalog No. NC0127657 Supplier Andwin Scientific Supplier No. 16466062
The 1.5ml SuperClear screwcap microtube feature a conical bottom, external graduations, universal screw threads and smooth surface for applying labels. It is constructed from ultra clear resin for easily viewing of contents. It includes attached natural low-profile caps with an ethylene propylene O-ring ensuring secure seal. They are compatible with most common rotors, autoclavable at 251F, freezable to -130F and radiation sterilized.