Learn More
Andwin Scientific TAPE ASSORTMENT1X500 12/CS
SDP
Catalog No. NC0148725
Encompass_Preferred
The Laboratory Labeling Tape can be utilized for several purposes, including general laboratory use. The tape sticks onto any surface and does not leave any residue. This product endures extreme temperatures from -23 to 121C (-10 to 250F). It has
Catalog No. NC0148725 Supplier Andwin Scientific Supplier No. 89098062
The Laboratory Labeling Tape can be utilized for several purposes, including general laboratory use. The tape sticks onto any surface and does not leave any residue. This product endures extreme temperatures from -23 to 121C (-10 to 250F). It has