Learn More
Andwin Scientific TAPE RAINBOW1/2X500"" 24/CS
SDP
Catalog No. NC0637725
Encompass_Preferred
The Laboratory Labeling Tape can be utilized for several purposes, including general laboratory use. The tape sticks onto any surface and does not leave any residue. This product endures extreme temperatures from -23 to 121C (-10 to 250F). It has
Catalog No. NC0637725 Supplier Andwin Scientific Supplier No. 89097920
The Laboratory Labeling Tape can be utilized for several purposes, including general laboratory use. The tape sticks onto any surface and does not leave any residue. This product endures extreme temperatures from -23 to 121C (-10 to 250F). It has