Learn More
Andwin Scientific TIP PIPET LTS AEROL PK960
SDP
Catalog No. NC0125751
Encompass
The Signature™ Pipet Tips are compatible with Rainin LTS pipettors. This product features ultrafine tips that can eject with significantly less force compared to most conventional pipet tips. The tips are sterile, are guaranteed to be free of RNase,
This item is not returnable. View return policy
Catalog No. NC0125751 Supplier Andwin Scientific Supplier No. 82003196PK
The Signature™ Pipet Tips are compatible with Rainin LTS pipettors. This product features ultrafine tips that can eject with significantly less force compared to most conventional pipet tips. The tips are sterile, are guaranteed to be free of RNase,