Learn More
Andwin Scientific TIP PIPET LTS AEROSL PK576
SDP
Catalog No. NC0125752
Encompass_Preferred
The Signature™ Pipet Tips are compatible with Rainin LTS pipettors. This product features ultrafine tips that can eject with significantly less force compared to most conventional pipet tips. The tips are sterile, are guaranteed to be free of RNase,
Catalog No. NC0125752 Supplier Andwin Scientific Supplier No. 82003198PK
The Signature™ Pipet Tips are compatible with Rainin LTS pipettors. This product features ultrafine tips that can eject with significantly less force compared to most conventional pipet tips. The tips are sterile, are guaranteed to be free of RNase,