Learn More
Andwin Scientific TUBE CLTBORO 12X75 CS1000
SDP
Catalog No. NC0474116
Encompass
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef
Catalog No. NC0474116 Supplier Andwin Scientific Supplier No. 47729570
The Culture Tubes feature resistance against chemical attack and weathering. In order to remove contamination, increase its strength, and lessen the chance of breakage, each tube is heat-treated at temperatures exceeding 500C (932F) immediately bef