Learn More
Andwin Scientific Centrifuge Tubes Ultra High Performance 15ml centrifuge tubes, 50/pk
SDP
Catalog No. NC0242711
Encompass_Preferred
Centrifuge Tubes Ultra High Performance 15ml centrifuge tubes for light sensitive samples. Tubes are made of dark amber polypropylene to protect light sensitive samples. Printed labeling area. Sterile. Max RCF: 12500g. 50 tubes with attached caps/
Catalog No. NC0242711 Supplier Andwin Scientific Supplier No. 89079541
Centrifuge Tubes Ultra High Performance 15ml centrifuge tubes for light sensitive samples. Tubes are made of dark amber polypropylene to protect light sensitive samples. Printed labeling area. Sterile. Max RCF: 12500g. 50 tubes with attached caps/