Learn More
Andwin Scientific TUBE MICRO 1.5ML PK500
SDP
Catalog No. NC0485068
Encompass_Preferred
The SuperClear™ Screw Cap Microcentrifuge Tubes feature an ultra-clear resin that permits easy viewing of contents. This product has universal screw threads that are compatible with most popular brand screw caps. The tubes are freestanding and nonste
Catalog No. NC0485068 Supplier Andwin Scientific Supplier No. 16466030
The SuperClear™ Screw Cap Microcentrifuge Tubes feature an ultra-clear resin that permits easy viewing of contents. This product has universal screw threads that are compatible with most popular brand screw caps. The tubes are freestanding and nonste