Learn More
Andwin Scientific TUBE PP ST GRD 50ML 500CS
SDP
Catalog No. NC9872140
Encompass_Preferred
The 50 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox
Catalog No. NC9872140 Supplier Andwin Scientific Supplier No. 21008178
The 50 mL SuperClear™ Ultra-High Performance Centrifuge Tubes feature a thick-wall construction that permits high-speed operation. The tubes are also leakproof, providing a secure storage for samples. This product is free of RNase, DNase, and endotox