Learn More
Andwin Scientific VWR TUBE ROTATOR WITH 36 X 1.
SDP
Catalog No. NC9854190
Encompass_Preferred
Tube Rotator Tube Rotator supplied with 42 x 1.5/2.0ML rotisserie. 18rpm fixed rotation speed. Adjustable rotisserie allows for mixing in the horizontal or vertical plane. 120V., 1/EA
Catalog No. NC9854190 Supplier Andwin Scientific Supplier No. 13916822
Tube Rotator Tube Rotator supplied with 42 x 1.5/2.0ML rotisserie. 18rpm fixed rotation speed. Adjustable rotisserie allows for mixing in the horizontal or vertical plane. 120V., 1/EA