Learn More
Andwin Scientific VWR VIAL TOC 10PPB CLR 80CS
SDP
Catalog No. NC3806418
Encompass
TOC VIALS 10PPB TOC. These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.
Catalog No. NC3806418 Supplier Andwin Scientific Supplier No. 76423174
TOC VIALS 10PPB TOC. These glass vial are used primarily for the collection and testing of aqueous samples for Total Organic Carbon (TOC). Secondary uses include containment of extracts. Please refer to the applicable certificate of analysis for certified analytes and corresponding reporting limits.