Learn More
Andwin Scientific Analog Vortex Mixer, 120 V
SDP
Catalog No. NC0951645
Encompass_Preferred
Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.
Catalog No. NC0951645 Supplier Andwin Scientific Supplier No. 10153838
Analog Vortex Mixers offer variable analog speed control with continuous or touch mode operation. Control allows low rpm start up for gentle shaking or high speed mixing for vigorous vortexing of samples. Two modes of operation; continuous mode when using optional accessory attachments or touch mode which activates mixing when depressing the cup head.