Learn More
Selleck Chemical LLC VX-11e 5mg 896720-20-0 VTX-11e, Vertex-11e
SDP
Supplier:  Selleck Chemical LLC S77095MG
Encompass_Preferred
VX-11e (VTX-11e, Vertex-11e) is a potent, selective, and orally bioavailable ERK2 inhibitor with Ki of <2 nM, over 200-fold selective over other kinases tested. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5158