Learn More
Selleck Chemical LLC Tezacaftor(VX-661) 5mg 1152311-62-0
SDP
Supplier:  Selleck Chemical LLC S70595MG
Encompass_Preferred
Tezacaftor¿(VX-661) is a second F508del CFTR corrector and is believed to help CFTR protein reach the cell surface. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0477441