Learn More
Apexbio Technology LLC VX-661(Synonyms: Tezacaftor, VX661, VX-661, VX 661), 10mM (in 1mL DMSO), CAS: 1152311-62-0.
SDP
Catalog No. 5000060286
Encompass_Preferred
VX-661 (CAS 1152311-62-0) is a small-molecule CFTR corrector developed by Vertex Pharmaceuticals designed to restore trafficking and enhance cellular surface expression of the mutated F508del-CFTR protein By facilitating proper folding and transport of this commonly mutated cystic fibrosis transmembrane conductance regulator (CFTR) VX-661 increases CFTR-mediated chloride channel activity in vitro Preclinical and clinical research have explored VX-661 alone and in combination with ivacaftor demonstrating enhanced F508del-CFTR functionality and providing a promising strategy for addressing cystic fibrosis caused by the F508del mutation
Catalog No. 50-000-60286 Supplier Apexbio Technology LLC Supplier No. A26645.1
VX-661 (CAS 1152311-62-0) is a small-molecule CFTR corrector developed by Vertex Pharmaceuticals designed to restore trafficking and enhance cellular surface expression of the mutated F508del-CFTR protein By facilitating proper folding and transport of this commonly mutated cystic fibrosis transmembrane conductance regulator (CFTR) VX-661 increases CFTR-mediated chloride channel activity in vitro Preclinical and clinical research have explored VX-661 alone and in combination with ivacaftor demonstrating enhanced F508del-CFTR functionality and providing a promising strategy for addressing cystic fibrosis caused by the F508del mutation