Learn More
TARGETMOL CHEMICALS INC VX-702 10MG
SDP
Catalog No. 502577102
Encompass_Preferred
Also available in 25mg 50mg and bulk. Please contact Fisher for quotes. VX-702 is a highly specific p38α MAPK inhibitor 14-fold higher selectivity for the p38α than p38β. VX-702 is a small molecule investigational oral anti-cytokine therapy for treatment of inflammatory diseases specifically rheumatoid arthritis (RA). purity: 99%
Catalog No. 50-257-7102 Supplier TARGETMOL CHEMICALS INC Supplier No. T251310MG
Also available in 25mg 50mg and bulk. Please contact Fisher for quotes. VX-702 is a highly specific p38α MAPK inhibitor 14-fold higher selectivity for the p38α than p38β. VX-702 is a small molecule investigational oral anti-cytokine therapy for treatment of inflammatory diseases specifically rheumatoid arthritis (RA). purity: 99%