Learn More
Selleck Chemical LLC VX-702
SDP
Catalog No. 5000004322
Encompass_Preferred
VX-702 is a highly selective inhibitor of p38 MAPK 14-fold higher potency against the p38 versus p38
Catalog No. 50-000-04322 Supplier Selleck Chemical LLC Supplier No. S60051G
VX-702 is a highly selective inhibitor of p38 MAPK 14-fold higher potency against the p38 versus p38