Learn More
Apexbio Technology LLC VX-702 479543-46-9; 745833-23-2 200mg
SDP
Catalog No. 5000096174
Encompass_Preferred
VX-702 is a small-molecule inhibitor targeting p38 mitogen-activated protein kinase (MAPK) also known as MAPK14 It is designed to inhibit p38 MAPK thereby regulating cellular responses to cytokine signaling and various stressors VX-702 exerts its biological activity primarily through ATP-competitive inhibition In experimental models VX-702 demonstrates potent inhibition with reported IC50 values ranging from 4 to 20 nM Based on these pharmacological properties VX-702 holds research potential in the study of p38 MAPK-dependent pathways platelet storage lesions and renal excretion mechanisms
Catalog No. 50-000-96174 Supplier Apexbio Technology LLC Supplier No. A8687200
VX-702 is a small-molecule inhibitor targeting p38 mitogen-activated protein kinase (MAPK) also known as MAPK14 It is designed to inhibit p38 MAPK thereby regulating cellular responses to cytokine signaling and various stressors VX-702 exerts its biological activity primarily through ATP-competitive inhibition In experimental models VX-702 demonstrates potent inhibition with reported IC50 values ranging from 4 to 20 nM Based on these pharmacological properties VX-702 holds research potential in the study of p38 MAPK-dependent pathways platelet storage lesions and renal excretion mechanisms