Learn More
Medchemexpress LLC VX-702 50mg | 745833-23-2 | 50MG
SDP
Catalog No. 5000243894
Encompass_Preferred
VX-702 is a highly selective inhibitor of p38 MAPK 14-fold higher potency against the p38 versus p38 [1
Catalog No. 50-002-43894 Supplier Medchemexpress LLC Supplier No. HY1040150MG
VX-702 is a highly selective inhibitor of p38 MAPK 14-fold higher potency against the p38 versus p38 [1