Learn More
Medchemexpress LLC VX-702 | 745833-23-2 | 99.7% | 404.32 | 25 MG
SDP
Catalog No. 5000347538
Encompass_Preferred
VX-702 is a highly selective inhibitor of p38α MAPK, demonstrating 14-fold higher potency against p38α compared to p38β. It is intended for research use only and not for patient use.
  • Highly selective p38α MAPK inhibitor, exhibiting 14-fold higher potency against p38α than p38β.
  • Inhibits p38 activation in platelets at 1 μM concentration.
  • No effect on platelet aggregation induced by p38 MAPK agonists.
  • Inhibits cytokine production of IL-6, IL-1β, and TNFα.
  • Long half-life of 16 to 20 hours in vivo.
  • Predominantly cleared renally.
  • Comparable efficacy to methotrexate and prednisolone in inhibiting wrist joint erosion and inflammation.
Catalog No. 50-003-47538 Supplier Medchemexpress LLC Supplier No. 25MGHY10401
VX-702 is a highly selective inhibitor of p38α MAPK, demonstrating 14-fold higher potency against p38α compared to p38β. It is intended for research use only and not for patient use.
  • Highly selective p38α MAPK inhibitor, exhibiting 14-fold higher potency against p38α than p38β.
  • Inhibits p38 activation in platelets at 1 μM concentration.
  • No effect on platelet aggregation induced by p38 MAPK agonists.
  • Inhibits cytokine production of IL-6, IL-1β, and TNFα.
  • Long half-life of 16 to 20 hours in vivo.
  • Predominantly cleared renally.
  • Comparable efficacy to methotrexate and prednisolone in inhibiting wrist joint erosion and inflammation.