Learn More
Abcam V x -745, p38 alpha MAPK inhibitor, 10MG
Catalog No. 502676157
Encompass_Preferred
MW 436.3 Da. Potent, selective p38 alpha MAPK inhibitor (IC₅₀ = 10 nM), with 20-fold selectivity over p38 beta and 1000-fold selectivity over closely related kinases, ERK1 and JNK1-3.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-6157 Supplier Abcam Supplier No. AB23036710MG
MW 436.3 Da. Potent, selective p38 alpha MAPK inhibitor (IC₅₀ = 10 nM), with 20-fold selectivity over p38 beta and 1000-fold selectivity over closely related kinases, ERK1 and JNK1-3.
The product is subject to the following: Abcam Restricted Use Statement