Learn More
Selleck Chemical LLC Belnacasan (VX-765) 10mg 273404-37-8
SDP
Catalog No. 508389
Encompass_Preferred
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0.8 nM in a cell-free assay. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-838-9 Supplier Selleck Chemical LLC Supplier No. S222810MG
Belnacasan (VX-765) is a potent and selective inhibitor of caspase-1 with Ki of 0.8 nM in a cell-free assay. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.