Learn More
Apexbio Technology LLC VX-809 936727-05-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062516
Encompass_Preferred
VX-809 (CAS 936727-05-8) is a small molecule CFTR corrector that partially restores the functional expression of F508del-CFTR the most common mutation associated with cystic fibrosis (CF) VX-809 stabilizes the misfolded CFTR protein by interacting with its membrane-spanning domain 1 (MSD1) thus facilitating proper maturation and membrane localization In F508del-CFTR-expressing Fischer rat thyroid (FRT) cells VX-809 enhanced CFTR protein processing (EC50 0 1 M) and improved chloride transport (EC50 0 5 M) VX-809 is utilized predominantly as a research tool in CF studies to understand and restore CFTR function often in combination with CFTR potentiators
Catalog No. 50-000-62516 Supplier Apexbio Technology LLC Supplier No. A83515.1
VX-809 (CAS 936727-05-8) is a small molecule CFTR corrector that partially restores the functional expression of F508del-CFTR the most common mutation associated with cystic fibrosis (CF) VX-809 stabilizes the misfolded CFTR protein by interacting with its membrane-spanning domain 1 (MSD1) thus facilitating proper maturation and membrane localization In F508del-CFTR-expressing Fischer rat thyroid (FRT) cells VX-809 enhanced CFTR protein processing (EC50 0 1 M) and improved chloride transport (EC50 0 5 M) VX-809 is utilized predominantly as a research tool in CF studies to understand and restore CFTR function often in combination with CFTR potentiators