Learn More
Selleck Chemical LLC VX-809 (Lumacaftor) 50mg 936727-05-8 VRT 826809
SDP
Catalog No. 508316
Encompass_Preferred
VX-809 (Lumacaftor, VRT 826809) acts to correct CFTR mutations common in cystic fibrosis by increasing mutant CFTR (F508del-CFTR) maturation,EC50 of 0.1 ?M in fisher rat thyroid cells. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-831-6 Supplier Selleck Chemical LLC Supplier No. S156550MG
VX-809 (Lumacaftor, VRT 826809) acts to correct CFTR mutations common in cystic fibrosis by increasing mutant CFTR (F508del-CFTR) maturation,EC50 of 0.1 ?M in fisher rat thyroid cells. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.