Learn More
Selleck Chemical LLC VY-3-135-E1147-10MM/1ML
SDP
Catalog No. 503137090
Encompass_Preferred
VY-3-135 a potent ACSS2 inhibitor with IC50 of 44 3 85 nM potently inhibits ACSS2 dependent fatty acid metabolism but has no effect on gene expression in tumors
Catalog No. 50-313-7090 Supplier Selleck Chemical LLC Supplier No. E114710MM/1ML
VY-3-135 a potent ACSS2 inhibitor with IC50 of 44 3 85 nM potently inhibits ACSS2 dependent fatty acid metabolism but has no effect on gene expression in tumors