Learn More
Mac-Mod Vydac 208MS C8, 300 A, 5 um, 4.6 x 250 mm HPLC Column
SDP
Supplier:  Mac-Mod 208MS54
Encompass
Vydac MS columns have a proprietary surface treatment and bonding process that provides unique selectivity. Suitable for the analysis of small peptides to large intact, undenatured proteins300A pore size spherical silica. Excellent peak shape with without TFA. The improved selectivity for peptides on the Vydac MS columns results in better primary structure definition and easier identification of degradation products and other protein characteristics. Vydac 218MS and 238MS are used for simple enzymatic digests (< 12 proteins) or biomolecules (0-5kDa). Vydac 208MS is ideal for biomolecules of molecular weight 5-10kDa. Vydac 214MS has lower capacity than C18 or C8 and is suitable for hydrophobic proteins or when minimal organic solvent is required. It is recommended for biomolecules of > 10kDa molecular weight, undenatured intact proteins, antibodies, oligonucleotides, human growth hormone. Vydac 219MS has the lowest capacity and is highly selective.
Catalog No. 50-112-0576