Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Vytac™ Formalin Disposal System, Vytac

Catalog No. 22046360 Shop All Epredia Products
Change view
Click to view available options
Description:
Vytac; 1 Gal. (3.79L)
Vytac; 5 Gal. (18.9L)
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
22-046-360 Vytac; 5 Gal. (18.9L)
22-046-332 Vytac; 1 Gal. (3.79L)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-046-360 Supplier Epredia™ Supplier No. 5405
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Safely dispose of both 10% buffered and unbuffered formalin, including zinc formalin, using the Epredia™ Vytac™ Formalin Disposal System.

Disposal Solution:
  • Breaks down formalin into inert components
  • Neutralizes four parts formalin waste per one part Vytac
  • When used as directed, neutralizes more than 99% of the waste formalin collected
Includes:
  • Vytac (1 gal.)
  • Reservoir with lid
  • Measuring cup
  • Plastic stir rod
Note: Vytac GNF will neutralize gluteraldehyde

Specifications

Product Type Vytac
Description Vytac; 5 Gal. (18.9L)
Quantity 5 Gal. (18.9 L)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.