Learn More
Drg International Inc DRG VZV IgA ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3522R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgA-class antibodies to Varicella Zoster Virus (VZV) in human serum or plasma (EDTA or heparin plasma). This product is for Research Use Only. The DRG VZV IgA ELISA is a solid phase enzyme-linked immunosorbent assay (ELISA). Samples are diluted with Sample Diluent and additionally incubated with IgG-RF-Sorbent, containing hyper-immune anti-human IgG class antibody to eliminate competitive inhibition from specific IgG and to remove rheumatoid factors. This pretreatment avoids false results. Microtiter wells as a solid phase are coated with Varicella Zoster Virus antigen. Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7592