Learn More
Drg International Inc DRG VZV IgG ELISA RUO, 96 Well Kit
Supplier:  Drg International Inc EIA3523R
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Varizella Zoster-Virus in serum or plasma. This product is for Research Use Only. The DRG VZV IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with inactivated Varizella Zoster-Virus antigen (wild strain isolate). Diluted specimens and ready-for-use controls are pipetted into these wells.
Catalog No. 50-194-7593