Learn More
Sino Biological VZV VZV-gE Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40907V08H100
Encompass_Preferred
A DNA sequence encoding the Varicella-zoster virus (VZV) / HHV-3 gE / glycoprotein E (Q9J3M8) (Met1-Tyr538) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-2078