Learn More
Apexbio Technology LLC W-13 hydrochloride 88519-57-7 1mg
SDP
Catalog No. 5000089561
Encompass_Preferred
W-13 hydrochloride (CAS 88519-57-7) is a small molecule inhibitor that acts as an antagonist of calmodulin thereby modulating calmodulin-dependent signaling pathways and influencing intracellular calcium homeostasis Studies have demonstrated that W-13 hydrochloride suppresses the proliferation of human breast cancer cells resistant to tamoxifen indicating its utility in investigating mechanisms underlying cancer drug resistance This compound is widely employed in biomedical research to dissect calmodulin-mediated signaling and to explore pathways involved in the adaptive responses of tumor cells
Catalog No. 50-000-89561 Supplier Apexbio Technology LLC Supplier No. B62761
W-13 hydrochloride (CAS 88519-57-7) is a small molecule inhibitor that acts as an antagonist of calmodulin thereby modulating calmodulin-dependent signaling pathways and influencing intracellular calcium homeostasis Studies have demonstrated that W-13 hydrochloride suppresses the proliferation of human breast cancer cells resistant to tamoxifen indicating its utility in investigating mechanisms underlying cancer drug resistance This compound is widely employed in biomedical research to dissect calmodulin-mediated signaling and to explore pathways involved in the adaptive responses of tumor cells