Learn More
Cayman Chemical W-13hydrochlorIde 10mg
SDP
Catalog No. 502954482
Encompass_Preferred
A potent antagonist of calmodulin (IC50 = 22 μM) that is widely used to investigate Ca2+/calmodulin-regulated enzyme activities
Catalog No. 50-295-4482 Supplier Cayman Chemical Supplier No. 1427710 MG
A potent antagonist of calmodulin (IC50 = 22 μM) that is widely used to investigate Ca2+/calmodulin-regulated enzyme activities