Learn More
Cayman Chemical W-3 ArachIdonIc AcId-d8 100ug
SDP
Catalog No. 502917733
Encompass_Preferred
An internal standard for the quantification of ω-3 arachidonic acid by GC- or LC-MS
Catalog No. 50-291-7733 Supplier Cayman Chemical Supplier No. 390011100 UG
An internal standard for the quantification of ω-3 arachidonic acid by GC- or LC-MS