Learn More
Enzo Life Sciences W-5 (20mg). CAS: 61714-25-8
SDP
Supplier:  Enzo Life Sciences BMLCA3150020
Encompass_Preferred
Binds to calmodulin, inhibiting Ca2+-calmodulin-regulated enzyme activity. Cell permeable. Alternative name: N-(6-Aminohexyl)-1-naphthalenesulfonamide . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water, DMSO (20mg/ml), 100% ethanol (hot) or methanol (hot). Long Term Storage: Ambient.
Catalog No. 50-201-0386